Tlačiť
Nitrile gloves, Omega™ Pro-Tech
Dodávateľ: NITRITEX
OPT2101-XS
NITROPT2101-XSEA 0 EUR
NITROPT2101-XS
Nitrile gloves, Omega™ Pro-Tech
Rukavice
These nitrile gloves feature an ambidextrous shape and beaded cuffs. They are resistant to a wide range of chemicals.
  • Non sterile, powder-free
  • Contain no natural latex proteins
  • Textured for improved grip
  • Beaded cuff provides easier donning and stability on the arm
  • Each size carries a unique bar code on the inner dispenser box (EAN-13) and the outer carton (ITF-14), which allows automatic data entry into stock control systems
EN 455 (Parts 1, 2 & 3), AQL 1.5
Certifikácie: EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010; Cat. III; AQL 1,5
Food contact approved