Tlačiť
Cleanroom gloves, latex, BioClean Legend™
Dodávateľ: NITRITEX
BLHN65 BLHN70 BLHN75 BLHN80 BLHN85 BLHN90
NITRBLHN65EA 393.75 EUR
NITRBLHN65 NITRBLHN70 NITRBLHN75 NITRBLHN80 NITRBLHN85 NITRBLHN90
Cleanroom gloves, latex, BioClean Legend™
Rukavice
These hand-specific cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments.
  • Protein level less than 50 µg/g
  • Textured finish for improved grip
  • Latex provides exceptional flexibility and comfort
  • Beaded cuff provides easier donning and stability on the arm
  • Anatomically shaped for enhanced wearer comfort
  • Each size carries a unique bar code on the carton (ITF-14), which allows automatic data entry into stock control systems
  • Cuff marking: Ink Print Size and L or R
  • Glove cleanliness LPC: <2000
  • Standard compliance: PPE,
More Product Information