Tlačiť
Utility tray
Dodávateľ: NEW PIG
PAK922
NEWPPAK922EA 126.98 EUR
NEWPPAK922
Utility tray
Podnosy/Misky/Kelímky Podnosy ochranné proti rozlití
This durable, one-piece utility trays contain liquid messes from oily parts and leaky containers.
  • Thick-wall, chemical-resistant polyethylene construction won't bend or warp
  • Ribs on bottom of tray add strength and elevate parts to keep them away from trapped liquids
  • Ideal for temporary storage of small containers that may leak or drip liquids onto the floor