Tlačiť
Seal for Screw Cap
Dodávateľ: NEUBERT VOLUME GLASSWAERE
6026-025-12
NEUB6026-025-12EA 11.54 EUR
NEUB6026-025-12
Seal for Screw Cap
Uzávěry Uzávěry k láhvím
Silicone with PTFE coating.
Seal for GL with hole for GL screw connections, silicone seal with hole.