Tlačiť
Fluid-transfer closure for bottles, Mason types, Nalgene™
Dodávateľ: Thermo Fisher Scientific
DS2153-0700
NALGDS2153-0700EA 72.5 EUR
NALGDS2153-0700
Fluid-transfer closure for bottles, Mason types, Nalgene™
Uzávěry Uzávěry k láhvím
PP, white.
  • External port tubes are moulded into closure for longer life
  • Port tubes accept ¼ in. flexible tubing
  • Vacuum limit: 127 mm Hg pressure limit: 0,137 bar
Fluid-transfer closure has a 70 mm closure that works with mason type bottles for fluid transfer in the lab or field. Can also connect with 125 and 500 ml of straight sided jars.