Tlačiť
Nalgene™ Screw Caps
Dodávateľ: Thermo Fisher Scientific
71-2240-1053 71-2150-0240 71-2150-0280 71-2150-0430 71-2150-0530 71-2150-0630 71-2150-1000 71-2151-0053 71-2151-0083 71-2151-0200 71-2151-0380 71-2154-0700 71-2160-0384 71-2160-0530 71-2160-0830 71-2171-0530 71-2172-0020 71-2174-0530 71-2401-0001 71-2401-0002 71-2411-1030 71-2411-1125 362150-0280 362150-4200 362150-5200 362150-6200 362150-1200 362150-1240 362150-0384 362150-0130 362150-0200 362150-0240 362150-1130 362150-1280 362150-1384 362150-4130 362150-4240 362150-4280 362150-4384 362150-5130 362150-5240 362150-5280 362150-5384 362150-6130 362150-6240 362150-6280 362150-6384 362150-7130 362150-7200 362150-7240 362150-7280 362150-7384
NALG71-2240-1053EA 215.5 EUR
NALG71-2240-1053 216-8953 216-8954 216-8957 216-8959 216-8960 216-8961 NALG71-2151-0053 NALG71-2151-0083 NALG71-2151-0200 NALG71-2151-0380 NALG71-2154-0700 NALG71-2160-0384 216-8972 216-8973 NALG71-2171-0530 NALG71-2172-0020 NALG71-2174-0530 NALG71-2401-0001 NALG71-2401-0002 NALG71-2411-1030 NALG71-2411-1125 215-1236 215-1250 215-1251 215-1252 215-1248 215-1249 215-1247 215-1591 215-1597 215-1598 215-1599 215-1600 215-1601 215-1602 215-1603 215-1604 215-1605 215-1606 215-1607 215-1608 215-1609 215-1610 215-1611 215-1612 215-1613 215-1618 215-1619 215-1620 215-1621 215-1622
Nalgene™ Screw Caps
Uzávěry Uzávěry k láhvím
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Certifikát ochrany životního prostředí IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement