Tlačiť
Drawer, VISION 17
Dodávateľ: MULTIROIR CONTROLEC
VISION17
MULCVISION17EA 266.37 EUR
MULCVISION17
Drawer, VISION 17
Krabice Boxy skladovací
This drawer block VISION 17 provide a clear and very compact storage of small parts of hardware.
  • Robust and shock resistant dark gray polypropylene body
  • Drawers are made of crystal polystyrene
W×D×H (interior): 200×93×93 mm
Weight: 1,64 kg