Tlačiť
Bunded spill flooring
Dodávateľ: MSG
8810 8811 8812 8813 8814 8815
MSGA8810EA 321.65 EUR
MSGA8810 MSGA8811 MSGA8812 MSGA8813 MSGA8814 MSGA8815
Bunded spill flooring
Absorpční materiály
The bunded spill flooring made from galavanised steel is designed for covering a large storage area.
  • Removable, hot-dip galvanized grids
  • Optimal adaption on space demands
  • Approved for the safe storage of water pollutants and flammable liquids
Load capacity: 5 000 kg/m²