Tlačiť
Face shields, V5 series
Dodávateľ: 3M
5A-CP
MMMA5A-CPEA 0 EUR
MMMA5A-CP
Face shields, V5 series
Štíty obličejové
Acetate face shields are lightweight and non-conductive.
  • Acetate visor
  • Lightweight
  • Non-conductive
3M™ headgear and face shields help provide head and eye protection against heat, debris, and chemicals. The 5A-CP offers chemical and impact resistance with its acetate face shield and its chin cup provides extended protection with downward visibility.