Tlačiť
Ultrafree®-CL Centrifugal Filter Units with Microporous Durapore® PVDF Membrane
Dodávateľ: MILLIPORE
UFC40HV25
MILYUFC40HV25EA 0 EUR
MILYUFC40HV25
Ultrafree®-CL Centrifugal Filter Units with Microporous Durapore® PVDF Membrane
Filtry Filtry centrifugační
These centrifugal filter units can be used for a number of applications, including removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.
  • Pre-sterilised units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 15 ml tubes
Hydrophilic PVDF membranes and PP housing.