Tlačiť
NovaMop™ Self Wringing Mops
Dodávateľ: MICRONOVA
QPNMSW-14
MICRQPNMSW-14EA 465.45 EUR
MICRQPNMSW-14
NovaMop™ Self Wringing Mops
Mopy
A flat head mop featuring a unique self-wringing mechanism that folds the mop head in half and draws upward for uniform wringing, every time. The 360° swivel joint allows full rotation of the mop head for use on floors, walls, and ceilings in the cleanroom.
  • Self wringing mop adapter with 360° swivel
  • One-step wringing process
  • Ideal, uniform wringing pressure
  • ACME thread adapter
  • Resistant to common cleaning chemicals and disinfectants
  • Autoclavable
The Micronova NovaMop™ Self Wringing Mop is designed for use with M-Zone® FlatMop™, SnapMop™, and PocketMop™More Product Information