Tlačiť
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Dodávateľ: Avantor Fluid Handling
MFLX96461-09EA 111.36 EUR
MFLX96461-09 MFLX96461-10 MFLX96461-12 MFLX96461-14 MFLX96461-16 MFLX96461-18 MFLX96461-19 MFLX96461-22 MFLX96461-24 MFLX96461-26 MFLX96461-27 MFLX96461-28 MFLX96461-29 MFLX96461-30 MFLX96461-31 MFLX96461-32 MFLX96461-34 MFLX96461-36 MFLX96461-38 MFLX96461-39 MFLX96461-40 MFLX96461-42 MFLX96461-44 MFLX96461-46 MFLX96461-48 MFLX96461-49
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Hadičky Hadičky peristaltické pumpy
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Ideal for general laboratory applications
  • Clear for easy sample viewing
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes inMore Product Information