Tlačiť
Masterflex® Sanitary Clamps, Nylon, Avantor®
Dodávateľ: Avantor Fluid Handling
MFLX31201-93EA 31.24 EUR
MFLX31201-93 MFLX31201-95 MFLX31201-97 MFLX31201-99
Masterflex® Sanitary Clamps, Nylon, Avantor®
Svorky Svorky trubkové
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance
  • Rated to 150 psi
  • Sterilizable via Autoclave and Gamma Irradiation
  • Features anti-tamper zip-tie lock out
  • Manufactured and packaged in cleanroom
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.