Tlačiť
CPC® Quick-Disconnect Fittings, Compression Bodies
Dodávateľ: Avantor Fluid Handling
06360-49 06360-50 06364-50 06364-50BLK 06360-50BLK 06360-49BLK
MFLX06359-90EA 22.43 EUR
MFLX06359-90 MFLX06359-91 MFLX06360-05 MFLX06360-98 MFLX06361-08 MFLX06364-90 MFLX06364-91 MFLX06360-49 MFLX06360-50 MFLX06364-50 MFLX06364-50BLK MFLX06360-50BLK MFLX06360-49BLK MFLX06360-05BLK MFLX06359-90BLK MFLX06364-90BLK MFLX06364-91BLK MFLX06360-98BLK MFLX06361-08BLK MFLX06359-91BLK MFLX06364-05BLK
CPC® Quick-Disconnect Fittings, Compression Bodies
Fitinky a konektory
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information