Tlačiť
Shelving systems, wire, Super Erecta® Pro™
Dodávateľ: METRO
PR1872NK3 PR2442NK3 PR2454NK3 PR2460NK3 PR2472NK3 PR2424NK3 PR2436NK3 PR2448NK3 PR1860NK3 PR1842NK3 PR1824NK3
METOPR1872NK3EA 245.54 EUR
METOPR1872NK3 METOPR2442NK3 METOPR2454NK3 METOPR2460NK3 METOPR2472NK3 METOPR2424NK3 METOPR2436NK3 METOPR2448NK3 METOPR1860NK3 METOPR1842NK3 METOPR1824NK3
Shelving systems, wire, Super Erecta® Pro™
Nábytek kancelářský Police
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration. Metroseal 3 with microban provides corrosion resistant finish for wet or high humidity environments with a 12 year warranty against rust and corrosion. Microban antimicrobial is built into the finish to keep the product 'cleaner between cleanings'.
  • Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
  • Versatile construction: by using various
More Product Information