Tlačiť
Neg-50™ Frozen Section Medium
Dodávateľ: Epredia
6502B
MENZ6502BEA 0 EUR
MENZ6502B
Neg-50™ Frozen Section Medium
Barviva Microscopy and staining reagents
Yield high-quality sections down to –50 °C without chatter using Epredia™ Neg-50™ frozen section medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8,0)
  • Specially formulated to prevent tissue separation from the slide
This colourless, water-soluble frozen section medium yields high-quality sections down to –50 °C without chatter. It leaves no residue after staining, and is compatible with the Microm® Automatic Cryo Approach system.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable