Tlačiť
Presentation Folder (for Cutting Tools)
Dodávateľ: MARTOR ARGENTAX
900852.08
MARO900852.08EA 0 EUR
MARO900852.08
Presentation Folder (for Cutting Tools)
Nože a řezačky
This folder is convient for large selection of cutting tools.
  • Dimension (W×D×H) : 40×250×330 mm
  • Basic material: Nylon
  • Weight product: 2,420 g
This folder is designed for transport cutting tools safely and present a convincing demonstration. Inside presentation folder contains high-quality safety knives and scissors at hand.
New Product