Tlačiť
Block heaters, NANOCOLOR®
Dodávateľ: MACHEREY-NAGEL
919350 919300
MANA919350EA 1701.58 EUR
MANA919350 MANA919300
Block heaters, NANOCOLOR®
Termostaty blokové Termostaty blokové/ohřívací
Digestion of the sample is an important step in photometric water analysis. The NANOCOLOR® VARIO 4 heating blocks with 24 holes for 16 mm Ø test tubes perform all required sample digestions in water and waste water analysis quickly and safely. The unit features 4 pre-programmed heating times: 30/60/120 min or continuous and has 8 free memory locations for individual heating times. There are 5 pre-programmed temperatures: 70/100/120/148/160 °C and 7 free memory locations for individual temperature settings. Temperature stability is ± 1 °C (according to DIN, EN, ISO and EPA methods). Warm-upMore Product Information