Tlačiť
TLC adsorbents, silica gel
Dodávateľ: MACHEREY-NAGEL
816320.1 816310.1 816310.5 816320.5 816330.1 816330.5 816340.1 816340.5 816380.1 816380.5 816400.1 816400.5 816410.1 816410.5
MANA816320.1EA 114.56 EUR
MANA816320.1 MANA816310.1 MANA816310.5 MANA816320.5 MANA816330.1 MANA816330.5 MANA816340.1 MANA816340.5 MANA816380.1 MANA816380.5 MANA816400.1 MANA816400.5 MANA816410.1 MANA816410.5
TLC adsorbents, silica gel
Chromatografii, Sorbenty Sorbenty pro TLC
Silica adsorbents for thin layer chromatography.
  • Available in different quality grades
  • Adsorbents with or without binder
  • Available with or without fluorescence indicator
Pore volume: 0,75 ml/g
Specific surface (BET): 500 m²/g