Tlačiť
Liquid chromatography sorbents, Polyamide 6
Dodávateľ: MACHEREY-NAGEL
815620.1 815600.1 815600.5 815610.1 815620.5 815610.5
MANA815620.1EA 580.01 EUR
MANA815620.1 MANA815600.1 MANA815600.5 MANA815610.1 MANA815620.5 MANA815610.5
Liquid chromatography sorbents, Polyamide 6
Chromatografii, Sorbenty HPLC sorbenty
Polyamide 6 adsorbents for column chromatography.
  • Polyamide 6 (ε-polycaprolactam)
  • Chromatographic quality
  • Separation mechanism mainly based on hydrogen bonds
Recommended application: separation of phenolic compounds (e.g., isolation of natural products), carboxylic acids and aromatic nitro compounds.