Tlačiť
VWR® Proteomics Grade, Tris-glycine buffer
Dodávateľ: VWR Chemicals
M115-40LEA 515.12 EUR
M115-40L
VWR® Proteomics Grade, Tris-glycine buffer
Pufry Pufry pro elektroforézu Pufry transferové
A pre-blended Tris-glycine powder for preparation of Laemmli SDS-PAGE running buffer. The 40 litre pack size prepares 40 litres of 1X Tris-glycine solution (0,025 M Tris base and 0,192 M glycine). Each Ready-Pack™ prepares 1 litre of a 10X concentrate.