Tlačiť
Orange Serum Agar
Dodávateľ: LIOFILCHEM
452690
LIOF452690EA 264.77 EUR
LIOF452690
Orange Serum Agar
Média mikrobiologická Připravená mikrobiologická média
Orange Serum Agar is a medium used for the isolation, cultivation and enumeration of microorganisms associated with spoilage of citrus fruits and derivates, such as lactic acid and acetic acid bacteria, yeasts and moulds.