Tlačiť
Perfection and Performance Laboratory Refrigerators with Stainless Steel Inner Liner, SRPvh & SRPvg
Dodávateľ: Liebherr
SRPVG6501 SRPVG6511 SRPVG8401 SRPVG8411 SRPVG1402 SRPVG1412 SRPVH6501 SRPVH6511 SRPVH8401 SRPVH8411 SRPVH1402 SRPVH1412
LIEBSRPVG6501EA 0 EUR
LIEBSRPVG6501 LIEBSRPVG6511 LIEBSRPVG8401 LIEBSRPVG8411 LIEBSRPVG1402 LIEBSRPVG1412 LIEBSRPVH6501 LIEBSRPVH6511 LIEBSRPVH8401 LIEBSRPVH8411 LIEBSRPVH1402 LIEBSRPVH1412
Perfection and Performance Laboratory Refrigerators with Stainless Steel Inner Liner, SRPvh & SRPvg
Chladničky
Glass/steel door high-volume refrigerators with forced air cooling. White steel exterior and stainless-steel interior. Choose between two temperature stability options to suit differing lab requirements. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer, and the LED interior lighting ensures good visibility of the contents of these large appliances. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals. These advanced lab fridges also feature a +2 °C safety device toMore Product Information