Tlačiť
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Dodávateľ: Liebherr
HMFVH4001 HMFVH4001H63 HMFVH4011 HMFVH4011H63 HMFVH5501 HMFVH5501H63 HMFVH5511 HMFVH5511H63
LIEBHMFVH4001EA 2643.3 EUR
LIEBHMFVH4001 LIEBHMFVH4001H63 LIEBHMFVH4011 LIEBHMFVH4011H63 LIEBHMFVH5501 LIEBHMFVH5501H63 LIEBHMFVH5511 LIEBHMFVH5511H63
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Chladničky
Ultra-efficient pharmaceutical refrigerators have been designed for the particular requirements with their dynamic cooling system, highly effective insulation, and precise temperature control, they guarantee reliable storage of sensitive substances and preparations.
  • Integrated data logger and USB interface for reading the data logger
  • PT1000 product sensor and Sensor feedthrough
  • Min./max. temperature log shows whether the storage temperatures were within the tolerance limits
  • Insulated and glass door models with vertical lighting, self-closing door at <90°
More Product Information