Tlačiť
Flasks, round bottom, wide neck, with beaded rim
Dodávateľ: LENZ LABORGLAS
07035028 07035037 07035049 07035058 07035070 07035073 07035079 07035088
LENZ07035028EA 149.81 EUR
LENZ07035028 LENZ07035037 LENZ07035049 LENZ07035058 LENZ07035070 LENZ07035073 LENZ07035079 LENZ07035088
Flasks, round bottom, wide neck, with beaded rim
Baňky Baňky s kulatým dnem
DURAN® Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • USP Standard
Marked with capacity and a white labelling area.
Certifikácie: DIN ISO 24450
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable