Tlačiť
Distilling bridges with splash head
Dodávateľ: LENZ LABORGLAS
05043002 05043003 05043004 05043001
LENZ05043002EA 138.99 EUR
LENZ05043002 LENZ05043003 271-3102 LENZ05043001
Distilling bridges with splash head
Komponenty pro destilaci Nástavce
DURAN®, borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.