Tlačiť
Adapters, NS socket to spherical joint ball
Dodávateľ: LENZ LABORGLAS
01531419 01532929 01532935 01532941
LENZ01531419EA 40.46 EUR
LENZ01531419 LENZ01532929 LENZ01532935 LENZ01532941
Adapters, NS socket to spherical joint ball
Adaptéry Adaptéry s kónickým zábrusů
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.