Tlačiť
Portable cart, Logic®
Dodávateľ: LABCONCO
8022000
LBCP8022000EA 0 EUR
LBCP8022000
Portable cart, Logic®
Vozíky
Handy cart stores lab supplies and may be moved easily to within reach of the user working at a bench, fume hood or biological safety cabinet. Three right side compartments accommodate pipettes and other supplies. Top left side compartment with front panel cutouts accommodates boxes of wipes and gloves. The hinged panel may be pivoted down for loading. Drawer holds a variety of miscellaneous small instruments and supplies. Open storage area in bottom holds larger equipment and supplies. Work surface and bottom are constructed of corrosion-resistant white high density polyethylene that isMore Product Information