Tlačiť
Accessories for microplate readers, µDrop Plate
Dodávateľ: THERMO LABSYSTEMS LIFE SCIENCE
N12391 N12391M2
736-0754EA 3359.5 EUR
736-0754 LABLN12391M2
Accessories for microplate readers, µDrop Plate
Čtečky mikrodestiček
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Up to 16 (µDrop Plate) or 32 (µDrop Duo Plate) sample positions for quick and easy measurement of sample volumes down to 2 μl
  • Compatible with an eight-channel pipette for sample addition
  • Easy to clean after measurement
  • Measurement slot for a rectangular cuvette
  • Ready-made Multiskan
More Product Information