Tlačiť

Zlato supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 60 nm OD1 (Abs. 540 nm)

Dodávateľ: Thermo Fisher Scientific

J67306.ACEA 123.77 EUR
J67306.AC
Zlato supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 60 nm OD1 (Abs. 540 nm)
Zlato

CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink Odour: Odourless MDL No.: MFCD00003436


Applications: For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Vzorec: Au
Mw: 196.97
Teplota varu: 2800
Teplota tavenia: 1063
Skladovacia teplota: Refrigerator
Číslo MDL: MFCD00003436
Číslo CAS: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR