Tlačiť
Steridon Sterile Coveralls with Hood
Dodávateľ: ISOFIELD IRELAND
80111M 80111S 80111XS 80111XXS 80111L 80111XL 80111XXL 80111XXXL
ISOF80111MEA 525.14 EUR
ISOF80111M ISOF80111S ISOF80111XS ISOF80111XXS ISOF80111L ISOF80111XL ISOF80111XXL ISOF80111XXXL
Steridon Sterile Coveralls with Hood
Jednorázové oděvy
The tri-laminate Steridon cleanroom coverall is aseptically folded and can be donned in 60 seconds or less.
  • Sterile cleanroom coverall with hood
  • Latex-free, enclosed and elasticated face, wrists, and ankles
  • Ergo-fit design at back and shoulders allowing greater range of motion
  • Aseptically presented fold
  • Colour-coded left and right inner tabs to facilitate donning
  • Removable external tabs at zipper to ensure sterile field is maintained
  • Validated for sterility by X-Ray
  • Sterility Assurance Level (SAL) 10⁻⁶
Steridon cleanroom coveralls are validated
More Product Information