Tlačiť
Homogeniser, HPH 2000/04
Dodávateľ: IKA JANKE KUNKEL
U066126 U071735 T061555 U068906
IKAAU066126EA 47591.83 EUR
IKAAU066126 IKAAU071735 IKAAT061555 IKAAU068906
Homogeniser, HPH 2000/04
Homogenizátory Homogenizátory stolní
The high pressure homogeniser HPH is an inline dispersing machine for energy-efficient, continuous production of superfine emulsions. Compared to other IKA inline machines, the dispersing action of the high pressure homogeniser HPH is not based on the rotor/stator principle. The improved homogenising effect is achieved within the homogeniser by means of spontaneous decompression of the liquid from max. 2000 bar to an ambient pressure. With the same energy input, the IKA HPH produces considerably finer emulsions than conventional toothed ring dispersers. The use of high pressure homogenisersMore Product Information