Tlačiť
Rotary Evaporator, RV 3 pro V-C Complete
Dodávateľ: IKA JANKE KUNKEL
10012324
IKAA10012324EA 11229.32 EUR
IKAA10012324
Rotary Evaporator, RV 3 pro V-C Complete
Odparky Odparky vakuové
The RV 3 pro V-C Complete package is the complete solution for your rotary evaporator setup as all components and connections required for operation are included.
  • Convenient package - everything that is needed to be ready for operation, from a single source
  • Intelligent laboratory equipment: All devices are perfectly coordinated and can even communicate with each other
  • High energy efficiency: Chiller and vacuum pump run only when necessary, saving valuable resources
The RV 3 rotary evaporator is the ideal entry-level model rotary evaporator. It isMore Product Information