Tlačiť
Homogeniser, FastPrep®-24, MP Biomedicals
Dodávateľ: MP BIOMEDICALS
116004500
ICNA116004500EA 11225.72 EUR
ICNA116004500
Homogeniser, FastPrep®-24, MP Biomedicals
Homogenizátory Homogenizátory stolní
FastPrep®-24 is used to homogenise biological samples. It lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
The FastPrep®-24 instrument is a high speed benchtop homogeniser offering the ultimate in speed and performance for the lysis of biological samples. Homogenisation of up to 24 samples takes place within 40 seconds. Uses a unique, optimised motion to disrupt cells through the multidirectional, simultaneous beating of specialised lysing matrix beads on the sample material.More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable