Tlačiť

L(+)-Kyselina askorbová sodná sůl 99, powder_white

Dodávateľ: MP Biomedicals

Synonymá: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

0210289025 0210289090 0210289091
ICNA0210289025EA 54.88 EUR
ICNA0210289025 ICNA0210289090 ICNA0210289091
L(+)-Kyselina askorbová sodná sůl 99, powder_white
L(+)-Kyselina askorbová sodná sůl

L-Ascorbic Acid, Sodium Salt is a vitamin widely distributed in the plant and animal kingdom.


  • pKa: pK1: 4.17, pK2: 11.57(Lit.)
  • Oxidizes on exposure to light and air


L-Ascorbic Acid, Sodium Salt is used as an antimicrobial and antioxidant in foodstuffs, a reductant for the isolation of chloroplasts. It has an effect on the nonspecific immune system. It preserves nitric oxide (NO) which may help prevent endothelial dysfunction, helps prevent and reverse the oxidative damage done to proteins and lipids as a result of cigarette smoke.


L-Ascorbic acid (Vitamin C) is a water soluble molecule use in a wide variety of applications, including cell culture, as a reducing agent that helps reduce oxidative stress. L-Ascorbate can be regenerated by biological systems.


Soluble in water (62 g/100 ml at 25 °C; 78 g/100 mL at 75 °C)

Vzorec: C₆H₇NaO₆
Mw: 198.11
Teplota tavenia: 213…222 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00082340
Číslo CAS: 134-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=1, water) +100° to +110°

ŠPECIFIKÁCIA

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR