Tlačiť
Stoppers, 20 mm, level 2 high-throughput applications, SureSTART™
Dodávateľ: Thermo Fisher Scientific
6A20BS 6A20BSF 6A20BST
HYPE6A20BSEA 25.56 EUR
HYPE6A20BS HYPE6A20BSF HYPE6A20BST
Stoppers, 20 mm, level 2 high-throughput applications, SureSTART™
Zátky Zátky kaučukové (gumové)
Use Thermo Scientific™ SureSTART™ 20 mm stoppers with crimp vials that have a 20 mm opening and a secure push fit, including our SureSTART 20 mm crimp headspace vials (level 2).
Select level 2 products for high throughput applications, when robustness and reproducibility are key. Choose from various pack sizes of low permeability, 3 mm thick bromobutyl rubber stoppers designed to provide an effective alternative to crimp caps.