Tlačiť
Two-dimensional BioProcess Container™ system, Labtainer™ BPC bags
Dodávateľ: THERMO BIOPROCESS DIVISION
SH30709.02 SH30712.04 SH30657.11 SH30657.13 SH30657.14 SH30657.15 SH30657.16 SH30658.11 SH30658.12 SH30658.13 SH30658.14 SH30658.15 SH30662.11 SH30658.16 SH30662.12 SH30662.14 SH30662.15 SH30662.16 SH30713.05 SH30714.05 SH30712.05 SH30713.01 SH30714.01 SH30714.02 SH30713.03 SH30712.02 SH30712.03 SH30713.04 SH30714.04 SH30709.01 SH30709.05 SH30713.02 SH30709.03
HYCPSH30709.02EA 107.35 EUR
HYCPSH30709.02 HYCPSH30712.04 HYCPSH30657.11 HYCPSH30657.13 HYCPSH30657.14 HYCPSH30657.15 HYCPSH30657.16 HYCPSH30658.11 HYCPSH30658.12 HYCPSH30658.13 HYCPSH30658.14 HYCPSH30658.15 HYCPSH30662.11 HYCPSH30658.16 HYCPSH30662.12 HYCPSH30662.14 HYCPSH30662.15 HYCPSH30662.16 HYCPSH30713.05 HYCPSH30714.05 HYCPSH30712.05 HYCPSH30713.01 HYCPSH30714.01 HYCPSH30714.02 HYCPSH30713.03 HYCPSH30712.02 HYCPSH30712.03 HYCPSH30713.04 HYCPSH30714.04 HYCPSH30709.01 HYCPSH30709.05 HYCPSH30713.02 HYCPSH30709.03
Two-dimensional BioProcess Container™ system, Labtainer™ BPC bags
Sáčky Sáčky na média
BioProcess Container™ (BPC™) systems include two-dimensional single-use bags for liquid handling. Manufactured from medical-grade plastics (cGMP compliant) and configured with ports, tubing and fittings, BPC™ systems provide an efficient means for sterile liquid handling in research and production settings.
  • Film type: CX5-14 Single Web
  • Sterilisation method: Gamma irradiation