Tlačiť
HyClone™ SFM4MAb media
Dodávateľ: HyClone products (Cytiva)
SH30513.02 SH30535.02
HYCLSH30513.02EA 102.94 EUR
HYCLSH30513.02 HYCLSH30535.02
HyClone™ SFM4MAb media
Média kultivační
HyClone™ SFM4MAb media was developed through the Metabolic Pathway Design approach to increase process yields for manufacturing of human and humanised recombinant antibodies for therapeutic use.