Tlačiť
Water baths, stainless steel for refrigeration
Dodávateľ: HUBER
2009.0032.00 2010.0026.00 2011.0022.00 2012.0026.00
HUBE2009.0032.00EA 4344.48 EUR
HUBE2009.0032.00 HUBE2010.0026.00 HUBE2011.0022.00 HUBE2012.0026.00
Water baths, stainless steel for refrigeration
Lázně Lázně vodní, nemíchané
The Cooling Bath are water baths with air-cooled refrigerating unit and use natural refrigerants. Housing and tank of stainless steel. The refrigerating unit works continuously.
In combination with an immersion circulator these refrigerated bath systems offers active cooling, in continuous operation over the complete working range.
Pozor: Cooling capacity at +20; 0; −10; −20; −30 °C.