Tlačiť
Cimarec™ VARIOMAG® i Telemodul Controller
Dodávateľ: THERMO H + P LABORTECHNIK
50119118
HPLT50119118EA 457.84 EUR
HPLT50119118
Cimarec™ VARIOMAG® i Telemodul Controller
Míchadla Nástroje na míchání
Save space with Thermo Scientific™ Cimarec™ i Telemodul controllers, including Micro, Mini, Compact and Maxi stirrers.
  • Provide stirring power for small to medium volumes of low-viscosity fluids
  • Recommended for single point stirring up to 250 ml
Objednávacie informácie: Order as replacement controller only.