Tlačiť
Self-Sealing Bags
Dodávateľ: HK PACK KRAUTSCHEID
102100 102120 102110 102210 102220 102230 102030 102010 102017 102011 102039 102040 102050 102060 102081 102090 102101 102200 102152 102542 102185 102123 102138 102036 102082 102070 102080_U
129-9136EA 52.07 EUR
129-9136 129-9137 129-9139 129-9151 129-9152 129-9153 HKPK102030 129-9140 129-9141 129-9149 129-9150 129-9142 129-9143 129-9144 129-9129 129-9146 129-9130 129-9148 HKPK102152 HKPK102542 HKPK102185 HKPK102123 HKPK102138 HKPK102036 HKPK102082 HKPK102070 HKPK102080_U
Self-Sealing Bags
Sáčky Sáčky skladovací
PE, 50 µm thick, transparent.
  • Available in several sizes
  • With or without printing area
These bags allow storage and transport of samples.