Tlačiť
Acetobacter Agar Dehydrated Culture Media
Dodávateľ: HIMEDIA
M238-500G
HIMEM238-500GEA 0 EUR
HIMEM238-500G
Acetobacter Agar Dehydrated Culture Media
Média mikrobiologická Mikrobiologická média dehydratovaná
Dehydrated culture media for enrichment, isolation, cultivation, and maintenance of microorganisms.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Application: Maintenance of glucose-positive Acetobacter species