Tlačiť
Avantor® Alltima HP, HPLC Columns, C18-HL, 5 µm
Dodávateľ: HICHROM
HICH87699EA 1278.19 EUR
HICH87699 HICH87700 HICH87697 HICH87698 HICH5135351 87624/N 87701/N
Avantor® Alltima HP, HPLC Columns, C18-HL, 5 µm
Kolony chromatografické
Avantor® Alltima HP columns contain the highest carbon load for superior retention and loadability.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
  • Low to no detectable column bleed
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.