Tlačiť
Avantor® Hichrom HI-VOC 2, GC Columns
Dodávateľ: Avantor
HI65-25-100-30EA 1224.51 EUR
HI65-25-100-30 HI65-25-100-60 HI65-25-150-30 HI65-25-150-60 HI65-25-200-30 HI65-25-200-60 HI65-25-300-30 HI65-25-300-60 HI65-32-100-105 HI65-32-100-30 HI65-32-100-60 HI65-32-100-75 HI65-32-150-105 HI65-32-150-30 HI65-32-150-60 HI65-32-150-75 HI65-32-200-105 HI65-32-200-30 HI65-32-200-60 HI65-32-200-75 HI65-32-2000-7.5 HI65-32-300-105 HI65-32-300-30 HI65-32-300-60 HI65-32-300-75 HI65-32-500-105 HI65-32-500-30 HI65-32-500-60 HI65-32-500-75 HI65-32-700-105 HI65-32-700-30 HI65-32-700-60 HI65-32-700-75 HI65-53-083-15 HI65-53-100-30 HI65-53-100-60 HI65-53-100-75 HI65-53-150-30 HI65-53-150-60 HI65-53-150-75 HI65-53-200-30 HI65-53-200-60 HI65-53-200-75 HI65-53-300-30 HI65-53-300-60 HI65-53-300-75 HI65-53-500-30 HI65-53-500-60 HI65-53-500-75 HI65-53-700-30 HI65-53-700-60 HI65-53-700-75 HIGCHI65-15-080-10 HIGCHI65-20-100-10 HIGCHI65-25-140-60
Avantor® Hichrom HI-VOC 2, GC Columns
Kolony chromatografické
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.