Tlačiť
CE capillaries, alignment interfaces and capillary cassette
Dodávateľ: AGILENT HEWLETT PACKARD
G7100-60002
HEWLG7100-60002EA 478.27 EUR
HEWLG7100-60002
CE capillaries, alignment interfaces and capillary cassette
Systémy elektroforetické Systémy pro Kapilární Klektroforézu
Alignment interfaces are an integral part of the Agilent diode-array detection (DAD) system. These interfaces contain optical slits which are precisely matched to the capillary inner diameter for optimised sensitivity and linear detection range.
In combination with the capillary cassette, alignment interfaces simplify capillary exchange, protect the fragile detection window and ensure exact alignment of the window in the detector. Quick-change cassette allows capillary exchange in less than one minute.
Cassette and interfaces accept all commercially available capillaries (~365 μm Ext.Ø)More Product Information