Tlačiť
Single element standard for icp-ms, yttrium
Dodávateľ: Agilent
5190-8586
HEWL5190-8586EA 112.16 EUR
HEWL5190-8586
Single element standard for icp-ms, yttrium
Standardy Standardy kationické Standardy - yttrium
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balenie: All standards packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.