Tlačiť
Capillaries for LC systems
Dodávateľ: AGILENT HEWLETT PACKARD
G1315-87311 5067-4660 5067-4647 5067-4649 5067-4650 5067-4651 5067-4653 5067-4670 5067-4658 5067-5107 5067-5108 5067-4669 5067-4735 5067-4744 5067-4746 5067-5104
HEWLG1315-87311EA 186.66 EUR
HEWLG1315-87311 HEWL5067-4660 HEWL5067-4647 HEWL5067-4649 HEWL5067-4650 HEWL5067-4651 HEWL5067-4653 HEWL5067-4670 HEWL5067-4658 HEWL5067-5107 HEWL5067-5108 HEWL5067-4669 HEWL5067-4735 HEWL5067-4744 HEWL5067-4746 HEWL5067-5104
Capillaries for LC systems
Hadičky Chromatography Tubing
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.