Tlačiť
Colour stripe silicone tubing, HelixMark®
Dodávateľ: HELIX MEDICAL EUROPE
61-079-27 61-080-21 61-080-23 61-080-26 61-080-27 61-080-30 61-080-32 61-079-23 61-079-26 61-079-30 61-079-32
HELI61-079-27EA 0 EUR
HELI61-079-27 HELI61-080-21 HELI61-080-23 HELI61-080-26 HELI61-080-27 HELI61-080-30 HELI61-080-32 HELI61-079-23 HELI61-079-26 HELI61-079-30 HELI61-079-32
Colour stripe silicone tubing, HelixMark®
Hadičky Přenosová hadička
Distinctive silicone colour striping enables users to recognise tube lines at a glance by application, tubing size, or functional area.
  • Fully autoclavable and sterilisable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification in every package
  • Lot traceability
Designed to suit a wide array of laboratory applications or pharmaceutical processes. All tubes provide exceptional biocompatibility, having undergone studies according to ISO 10993-1 and meeting USP Class VI requirements. TheMore Product Information