Tlačiť
Assistent Brushes for Flasks
Dodávateľ: GLASWARENFABRIK KARL HECHT
91120000017 91120000018 91120000019 91120000020 41912010
HECH91120000017EA 364.11 EUR
HECH91120000017 HECH91120000018 HECH91120000019 HECH91120000020 HECH41912010
Assistent Brushes for Flasks
Kartáče
Straight horsehair brushes with galvanized iron thread handle.