Tlačiť
Staining trough
Dodávateľ: GLASWARENFABRIK KARL HECHT
41201042
HECH41201042EA 173.04 EUR
HECH41201042
Staining trough
Nádoby Nádoby pro barvení
Staining system contains staining jar with ground-in cover with knob made of glass, rack and handle made from stainless steel.
Suitable for 26 mm wide microscopic slides. Ideal for serial stainings, especially for the method according to Papanicolaou.